Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNURF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566794 Supplier Invitrogen™ Supplier No. PA566794
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRRTTEQHVPE VEVQVKRRRT ASLSNQECQL YPRRS Highest antigen sequence identity to the following orthologs - mouse 97%, rat 94%.

SNURF is a highly basic protein localized to the nucleus. The evolutionarily constrained open reading frame of its gene is found on a bicistronic transcript which has a downstream ORF encoding the small nuclear ribonucleoprotein polypeptide N. The upstream coding region utilizes the first three exons of the transcript, a region that has been identified as an imprinting center. Multiple transcription initiation sites have been identified and extensive alternative splicing occurs in the 5' untranslated region but the full-length nature of these transcripts has not been determined. An alternate exon has been identified that substitutes for exon 4 and leads to a truncated, monocistronic transcript. Alternative splicing or deletion caused by a translocation event in the 5' UTR or coding region of this gene leads to Angelman syndrome or Prader-Willi syndrome due to parental imprint switch failure. The function of this protein is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNURF
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNURF
Gene Accession No. Q9Y675
Gene Alias HCERN3; Sm protein D; Sm protein N; Small nuclear ribonucleoprotein-associated protein N; Sm-D; SMN; Sm-N; snRNP-N; SNRPN; SNRPN upstream reading frame; SNRPN upstream reading frame protein; SNURF; Tissue-specific-splicing protein
Gene Symbols SNURF
Host Species Rabbit
Immunogen Recombinant Human SNURF. Recombinant protein control fragment (Product #RP-107452).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8926
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.