Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNW1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520308
Description
A suggested positive control is mouse skeletal muscle tissue lysate. PA5-20308 can be used with blocking peptide PEP-0402.
CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.
Specifications
| SNW1 | |
| Polyclonal | |
| Unconjugated | |
| Snw1 | |
| 2310008B08Rik; AW048543; Bx42; homolog of Drosophila BX42; NCOA-62; Nuclear protein SkiP; nuclear receptor coactivator NCoA-62; nuclear receptor coactivator, 62-kD; PPS; Prp45; PRPF45; RGD1561926; SKI interacting protein; ski-interacting protein; Skiip; SKIP; SNW domain containing 1; SNW domain-containing protein 1; SNW1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22938, 500695, 66354 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q13573, Q9CSN1 | |
| Snw1 | |
| A 16 amino acid peptide from near the carboxy terminus of human SkiP. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction