Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX1 Recombinant Rabbit Monoclonal Antibody (8G8Z2)
GREENER_CHOICE

Catalog No. PIMA538065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538065 Supplier Invitrogen™ Supplier No. MA538065
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, 293T, HepG2, Rat testis Immunogen sequence: MASGGGGCSA SERLPPPFPG LEPESEGAAG GSEPEAGDSD TEGEDIFTGA AVVSKHQSPK ITTSLLPINN GSKENGIHEE QDQEPQDLFA DATVELSLDS.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 8G8Z2
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SNX1
Gene Accession No. Q13596
Gene Alias HsT17379; MGC8664; Snx1; SNX1A; sorting nexin 1; sorting nexin 1A; sorting nexin-1; VPS5
Gene Symbols SNX1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human SNX1 (Q13596).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6642
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.