Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX10 Antibody (OTI6B7), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274262AF532
Description
SNX10 Monoclonal specifically detects SNX10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SNX10 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
SNX10 | |
Full length recombinant protein of human SNX10 (NP_037454) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI6B7 | |
Western Blot, Immunohistochemistry | |
MGC33054, sorting nexin 10, sorting nexin-10 | |
Mouse | |
0.1 mL | |
29887 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction