Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX10 Antibody (OTI6B7), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274262JF549
Description
SNX10 Monoclonal specifically detects SNX10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SNX10 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SNX10 | |
Full length recombinant protein of human SNX10 (NP_037454) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI6B7 | |
Western Blot, Immunohistochemistry | |
MGC33054, sorting nexin 10, sorting nexin-10 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
29887.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction