Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX11 Monoclonal Antibody (2G1)
GREENER_CHOICE

Catalog No. PIMA521788
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521788 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521788 Supplier Invitrogen™ Supplier No. MA521788
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MGFWCRMSEN QEQEEVITVR VQDPRVQNEG SWNSYVDYKI FLHTNSKAFT AKTSCVRRRY REFVWLRKQL QRNAGLVPVP ELPGKSTFFG TSDEFIEKRR QGLQHFLEKV LQSVVLLSDS QLHLFLQSQL SVPEIEACVQ GRSTMTVSDA ILRYAMSNCG WAQEERQSSS HLAKGDQPKS CCFLPRSGRR SSPSPPPSEE KDHLEVWAPV VDSEVPSLES PTLPPLSSPL CCDFGRPKEG TSTLQSVRRA VGGDHAVPLD PGQLETVLEK.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members. This gene encodes a protein of unknown function. This gene results in two transcript variants differing in the 5' UTR, but encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX11
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2G1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SNX11
Gene Accession No. Q9Y5W9
Gene Alias 4933439F10Rik; A930041K09Rik; AI117694; MGC111019; SNX11; sorting nexin 11; sorting nexin-11
Gene Symbols SNX11
Host Species Mouse
Immunogen SNX11 (AAH00768, 1 a.a. ~ 270 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29916
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.