Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560827 Supplier Invitrogen™ Supplier No. PA560827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LFLQSQLSVP EIEACVQGRS TMTVSDAILR YAMSNCGWAQ EERQSSSHLA KGDQPKSCCF LPRSGRRSSP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members. This gene encodes a protein of unknown function. This gene results in two transcript variants differing in the 5' UTR, but encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX11
Gene Accession No. Q91WL6, Q9Y5W9
Gene Alias 4933439F10Rik; A930041K09Rik; AI117694; MGC111019; SNX11; sorting nexin 11; sorting nexin-11
Gene Symbols SNX11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX11. Recombinant protein control fragment (Product #RP-100378).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29916, 303493, 74479
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.