Learn More
Invitrogen™ SNX12 Monoclonal Antibody (OTI7B3)

Description
SNX12 Monoclonal Antibody for IHC (P), Western Blot
Specifications
Specifications
| Antigen | SNX12 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI7B3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SNX12 |
| Gene Accession No. | Q9UMY4 |
| Gene Alias | 2610001F05Rik; AW045757; fc02c04; fc03c06; fj97e07; LOW QUALITY PROTEIN: sorting nexin-12; MGC118982; MGC118983; RGD1565585; SDP8; SDP8 protein; similar to sorting nexin 12; snx12; sorting nexin 12; sorting nexin 12, variation 1; sorting nexin 12, variation 2; sorting nexin-12; wu:fc02c04; wu:fc03c06; wu:fj97e07; zgc:158590; zgc:55728; zgc:77034; zgc:77034 protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.