Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNX12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599046
Description
SNX12 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members. A similar protein in mouse may be involved in regulating the neurite outgrowth. Alternate splicing results in multiple transcript variants.
Specifications
SNX12 | |
Polyclonal | |
Unconjugated | |
SNX12 | |
2610001F05Rik; AW045757; fc02c04; fc03c06; fj97e07; LOW QUALITY PROTEIN: sorting nexin-12; MGC118982; MGC118983; RGD1565585; SDP8; SDP8 protein; similar to sorting nexin 12; snx12; sorting nexin 12; sorting nexin 12, variation 1; sorting nexin 12, variation 2; sorting nexin-12; wu:fc02c04; wu:fc03c06; wu:fj97e07; zgc:158590; zgc:55728; zgc:77034; zgc:77034 protein | |
Rabbit | |
Protein G | |
RUO | |
29934 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1.0 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9UMY4 | |
SNX12 | |
Recombinant Human Sorting nexin-12 protein (2-172AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction