Learn More
Invitrogen™ SNX12 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
Specifications
Specifications
| Antigen | SNX12 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | SNX12 |
| Gene Accession No. | Q9UMY4 |
| Gene Alias | 2610001F05Rik; AW045757; fc02c04; fc03c06; fj97e07; LOW QUALITY PROTEIN: sorting nexin-12; MGC118982; MGC118983; RGD1565585; SDP8; SDP8 protein; similar to sorting nexin 12; snx12; sorting nexin 12; sorting nexin 12, variation 1; sorting nexin 12, variation 2; sorting nexin-12; wu:fc02c04; wu:fc03c06; wu:fj97e07; zgc:158590; zgc:55728; zgc:77034; zgc:77034 protein |
| Gene Symbols | SNX12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.