Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588780 Supplier Invitrogen™ Supplier No. PA588780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FGKTNSEKYL EQCEHSFLPP TSPGVPKCLE EMKREARTIK IDRRLTGANI IDEPLQQVIQ FSLRDYVQYW YYTLSDDESF LLEIRQTLQN ALIQFATRSK EIDWQPYFTT RIVDDFGTHL RVFRKAQQKI TEKDDQVKGT AEDLVDTFFE VEVEMEKEVC RDLVCTSPKD EEGFLRDLCE V; Positive Samples: U-87MG, HeLa; Cellular Location: Cytoplasmic side, Early endosome membrane, Peripheral membrane protein.

May be involved in several stages of intracellular trafficking. May play a role in endosome homeostasis. Acts as a GAP for Galphas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX13
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SNX13
Gene Accession No. Q6PHS6, Q9Y5W8
Gene Alias KIAA0713; mKIAA0713; RGS domain- and PHOX domain-containing protein; RGS-PX1; Snx13; sorting nexin 13; sorting nexin-13
Gene Symbols SNX13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-230 of human SNX13 (NP_0559471).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217463, 23161
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.