Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNX13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119427
This item is not returnable.
View return policy
Description
SNX13 Polyclonal Antibody for Western Blot, IHC (P)
May be involved in several stages of intracellular trafficking. May play a role in endosome homeostasis. Acts as a GAP for Galphas.
Specifications
SNX13 | |
Polyclonal | |
Unconjugated | |
SNX13 | |
KIAA0713; mKIAA0713; RGS domain- and PHOX domain-containing protein; RGS-PX1; Snx13; sorting nexin 13; sorting nexin-13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23161 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9Y5W8 | |
SNX13 | |
Recombinant protein Sorting Nexin 13. The antigen corresponds to amino acid range 97-284 of the target protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction