Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553615
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553615 Supplier Invitrogen™ Supplier No. PA553615
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LFRFMNFLKQ EGAVHVLQFC LTVEEFNDRI LRPELSNDEM LSLHEELQKI YKTYCLDESI DKIRFDPFIV EEIQRIAEGP YIDVVKLQTM RCLFEAYEHV LSLLENVFTP MFCHSDEYFR QLLRGAESPT RN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Plays a role in maintaining normal neuronal excitability and synaptic transmission. May be involved in several stages of intracellular trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Snx14
Gene Accession No. Q8BHY8, Q9Y5W7
Gene Alias B830022K16; C330035N22Rik; RGS-PX2; SCAR20; SNX14; sorting nexin 14; sorting nexin-14; YR-14
Gene Symbols Snx14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX14. Recombinant protein control fragment (Product #RP-90962).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 244962, 315871, 57231
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.