Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX15 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001210
Description
SNX15 Polyclonal specifically detects SNX15 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
SNX15 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
clone iota unknown protein, HSAF001435, sorting nexin 15, sorting nexin-15 | |
Goat | |
Affinity Purified | |
RUO | |
29907 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1:1000, Peptide ELISA Detection limit 1:8000 | |
NP_037438 | |
SNX15 | |
Peptide with sequence C-EEILRLHLSQLPP corresponding to C-Terminus according to NP_037438.2, NP_680086.2. | |
0.1 mg | |
Primary | |
This antibody is expected to recognize both Human isoforms according to NP_037438.2 and NP_680086.2. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction