Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110409
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110409 Supplier Invitrogen™ Supplier No. PA5110409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSRQAKDDFL RHYTVSDPRT HPKGYTEYKV TAQFISKKDP EDVKEVVVWK RYSDFRKLHG DLAYTHRNLF RRLEEFPAFP RAQVFGRFEA SVIEERRKGA EDLLRFTVHI PALNNSPQLK EFFRGGEVTR PLEVSRDLHI LPPPLIPTPP PDDPRLSQLL PAERRGLEEL EVPVDPPPSS PAQEALDLLF NCESTEEASG SPARGPLTEA ELALFDPFSK EEGAAPSPTH VAELATMEVE SARLDQEPWE PGGQEEEEDG EGGPTPAYLS QATELITQAL RDEKAGAYAA ALQGYRDGVH VLLQGVPSDP LPARQEGVKK KAAEYLKRAE EILRLHLSQL PP.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Overexpression of this gene results in a decrease in the processing of insulin and hepatocyte growth factor receptors to their mature subunits. This decrease is caused by the mislocalization of furin, the endoprotease responsible for cleavage of insulin and hepatocyte growth factor receptors. This protein is involved in endosomal trafficking from the plasma membrane to recycling endosomes or the trans-Golgi network. Alternative splicing results in multiple transcript variants. Read-through transcription also exists between this gene and the upstream ADP-ribosylation factor-like 2 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX15
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Snx15
Gene Accession No. Q4V896, Q91WE1, Q9NRS6
Gene Alias 1500032B08Rik; clone iota unknown protein; E130013C21Rik; hCG_23393; HSAF001435; OTTHUMP00000035515; SNX15; sorting nexin 15; sorting nexin-15
Gene Symbols Snx15
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-342 of human SNX15 (NP_037438.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293691, 29907, 69024
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.