Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX16 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318461AF488
Description
SNX16 Polyclonal antibody specifically detects SNX16 in Human samples. It is validated for Western BlotSpecifications
SNX16 | |
Polyclonal | |
Western Blot | |
DKFZp666H147, sorting nexin 16, sorting nexin-16 | |
Recognizes region between residues Between Between 250 and 300.. | |
0.1 mL | |
Signal Transduction | |
64089 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction