Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555165 Supplier Invitrogen™ Supplier No. PA555165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FPGFRLALPP KRWFKDNYNA DFLEDRQLGL QAFLQNLVAH KDIANCLAVR EFLCLDDPPG PFDSLEESRA FCETLEETNY RLQKELLEKQ KEMESL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. The function of this protein has not been determined. This gene results in three transcript variants encoding two distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX16
Gene Accession No. P57768, P57769, Q8C080
Gene Alias 4930522N22Rik; AI117577; AI747675; DKFZp666H147; Snx16; sorting nexin 16; sorting nexin-16
Gene Symbols SNX16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX16. Recombinant protein control fragment (Product #RP-93921).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64088, 64089, 74718
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.