Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX16 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP261225
Description
SNX16 Polyclonal specifically detects SNX16 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
SNX16 | |
Polyclonal | |
Western Blot 1:1000, Immunoprecipitation 20 ul/mg lysate | |
P57768 | |
SNX16 | |
100% Sequence Identity Orangutan between 250 and 300 | |
0.1 mL | |
64089 | |
Store the unopened product at 2 - 8degrees C. Protect from light. Do not use past expiration date. |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
DKFZp666H147, sorting nexin 16, sorting nexin-16 | |
Rabbit | |
Affinity Purified | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction