Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587768 Supplier Invitrogen™ Supplier No. PA587768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MHFSIPETES RSGDSGGSAY VAYNIHVNGV LHCRVRYSQL LGLHEQLRKE YGANVLPAFP PKKLFSLTPA EVEQRREQLE KYMQAVRQDP LLGSSETFNS FLRRAQQETQ QVPTEEVSLE VLLSNGQKVL VNVLTSDQTE DVLEAVAAKL DLPDDLIGYF SLFLVREKED GAFSFVRKLQ EFELPYVSVT SLRSQEYKIV LRKSYWDSAY DDDVMENRVG LNLLYAQTVS DIERGWILVT KEQHRQLKSL QEKVSKKEFL RLAQTLRHYG; Positive Samples: A-549; Cellular Location: Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle membrane, Early endosome, Peripheral membrane protein.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a B41 domain. This protein interacts with the cytoplasmic domain of P-selectin, and may function in the intracellular trafficking of P-selectin. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX17
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Snx17
Gene Accession No. Q15036
Gene Alias 5830447M19Rik; AI790646; b2b1625.1Clo; D5Ertd260e; KIAA0064; mKIAA0064; Snx17; sorting nexin 17; sorting nexin-17
Gene Symbols Snx17
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human SNX17 (NP_0555631).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9784
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.