Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106319 Supplier Thermo Scientific Supplier No. PA5106319
Add to Cart
Add to Cart

SNX17 Polyclonal Antibody

SNX17 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX17
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Snx17
Gene Accession No. Q15036, Q6AYS6, Q8BVL3
Gene Alias 5830447M19Rik; AI790646; b2b1625.1Clo; D5Ertd260e; KIAA0064; mKIAA0064; Snx17; sorting nexin 17; sorting nexin-17
Gene Symbols Snx17
Host Species Rabbit
Immunogen A synthesized peptide derived from human SNX17(Accession Q15036), corresponding to amino acid residues S66-E116.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266781, 298836, 9784
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.