Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558113
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558113 Supplier Invitrogen™ Supplier No. PA558113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDEWDDSSTV ADEPGALGSG AYPDLDGSSS AGVGAAGRYR LSTRSDLSLG SRGGSVPPQH HPSGPKSSAT VSRNLN Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a SH3 domain. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX18
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX18
Gene Accession No. Q91ZR2, Q96RF0
Gene Alias SH3 and PX domain-containing protein 3B; SH3PX2; SH3PXD3B; Snag1; Snx18; sorting nexin 18; sorting nexin associated golgi protein 1; sorting nexin-18; Sorting nexin-associated Golgi protein 1
Gene Symbols SNX18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX18. Recombinant protein control fragment (Product #RP-96965).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112574, 170625, 310097
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.