Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX2 Antibody, DyLight 680 , Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224514FR
Description
SNX2 Polyclonal antibody specifically detects SNX2 in Human, Mouse, Rat, Primate samples. It is validated for Western BlotSpecifications
SNX2 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate | |
Purified |
Western Blot | |
DyLight 680 | |
MGC5204, sorting nexin 2, sorting nexin-2, Transformation-related gene 9 protein, TRG-9 | |
A portion of amino acids 120-170 of human Snx2 was used as the immunogen. | |
0.1 mL | |
Signal Transduction | |
6643 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction