Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen SNX2
Concentration 0.5 mg/mL
Dilution Western Blot 0.25-0.50 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224514SS
SDP
View Documents
Novus Biologicals
NBP224514SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224514
SDP
View Documents
Novus Biologicals
NBP224514
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

SNX2 Polyclonal specifically detects SNX2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot.
Specifications

Specifications

SNX2
Western Blot 0.25-0.50 ug/ml
Polyclonal
Rabbit
Signal Transduction
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
6643
A portion of amino acids 120-170 of human Snx2 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
RUO
Human, Mouse, Rat, Primate
MGC5204, sorting nexin 2, sorting nexin-2, Transformation-related gene 9 protein, TRG-9
SNX2
IgG
Affinity Purified
The amino acid sequence used as immunogen is 100% homologous in human, mouse and bovine, and 77% homologous in chicken.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.