Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32143025UL
Description
SNX2 Polyclonal antibody specifically detects SNX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SNX2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
MGC5204, sorting nexin 2, sorting nexin-2, Transformation-related gene 9 protein, TRG-9 | |
Produced in rabbits immunized with E. coli-derived Human SNX2 fragment | |
25 μg | |
Signal Transduction | |
6643 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction