Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145171
Description
Immunogen sequence: SPSSPEPASL PAEDISANSN GPKPTEVVLD DDREDL Antigen sequence identity: Mouse 94%; Rat 89%.
May be involved in several stages of intracellular trafficking.
Specifications
SNX2 | |
Polyclonal | |
Unconjugated | |
SNX2 | |
0610030A03Rik; CTB-36H16.2; hypothetical protein LOC509769; MGC5204; SNX2; sorting nexin 2; sorting nexin-2; Transformation-related gene 9 protein; TRG9; TRG-9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
6643 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60749 | |
SNX2 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 38-73. Recombinant protein control fragment (Product #RP-109875). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction