Learn More
SNX2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304544AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein associates with formin-binding protein 17, but its function is unknown. This protein may form oligomeric complexes with family members.Specifications
SNX2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O60749, Q9CWK8 | |
SNX2 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SNX2 | |
MGC5204, TRG-9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6643, 67804 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.