Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNX20 Monoclonal Antibody (4E9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA522986
Description
Peptide Sequence: SGLSSNSSMT TRELQQYWQN QKCRWKHVKL LFEIASARIE ERKVSKFVMG KSRPGEMTYP GSRGETGTAP EPDPRCPRQS DM.
SNX20 interacts with the cytoplasmic domain of PSGL1 and cycles PSGL1 into endosomes.
Specifications
SNX20 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q7Z614 | |
SNX20 | |
SNX20 (NP_699168.1, 47 a.a. ~ 128 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry | |
4E9 | |
Unconjugated | |
SNX20 | |
9130017C17Rik; RGD1307787; selectin ligand interactor cytoplasmic 1; selectin ligand interactor cytoplasmic-1; selectin ligand-interactor cytoplasmic 1; SLIC1; SLIC-1; SNX20; sorting nexin 20; sorting nexin-20 | |
Mouse | |
Affinity chromatography | |
RUO | |
124460 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction