Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNX20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112719
Description
SNX20 Polyclonal Antibody for Western Blot, IHC (P), ELISA
SNX20 interacts with the cytoplasmic domain of PSGL1 and cycles PSGL1 into endosomes.
Specifications
| SNX20 | |
| Polyclonal | |
| Unconjugated | |
| SNX20 | |
| 9130017C17Rik; RGD1307787; selectin ligand interactor cytoplasmic 1; selectin ligand interactor cytoplasmic-1; selectin ligand-interactor cytoplasmic 1; SLIC1; SLIC-1; SNX20; sorting nexin 20; sorting nexin-20 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 124460, 71607 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q7Z614, Q9D2Y5 | |
| SNX20 | |
| Recombinant Human Sorting nexin-20 protein (1-102AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction