Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 131801AP150UL
Description
SNX20 interacts with the cytoplasmic domain of PSGL1 and cycles PSGL1 into endosomes.Specifications
| SNX20 | |
| Polyclonal | |
| Unconjugated | |
| SNX20 | |
| SLIC 1, SLIC1, SNX20, sorting nexin 20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 124460, 307742, 71607 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5BK61, Q7Z614, Q9D2Y5 | |
| Snx20 | |
| SNX20 Fusion Protein Ag3847 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction