Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX21 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298062G
Description
SNX21 Polyclonal specifically detects SNX21 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SNX21 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C20orf161, chromosome 20 open reading frame 161, MGC29895, PP3993, SNXL, SNX-LdJ337O18.4, sorting nexin 21, sorting nexin family member 21, Sorting nexin L, sorting nexin-21 | |
Produced in rabbits immunized with E. coli-derived Human SNX21 fragment. | |
0.1 mL | |
Signal Transduction | |
90203 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction