Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558407 Supplier Invitrogen™ Supplier No. PA558407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLEAYIQGIL YLNQEVPKEL LEFLRLRHFP TDPKASNWGT LREFLPGDSS SQQHQRPVLS FHVDPYVCNP SPESLPNVVV NGVLQGLYSF SISPDKAQPK AACHPAPL Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 65%.

The protein encoded by this gene is a sorting nexin that is found in the cytoplasm, where it interacts with membrane-bound phosphatidylinositol 3-phosphate. The encoded protein may play a role in intracellular trafficking. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX22
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX22
Gene Accession No. Q96L94
Gene Alias Gm1120; SNX22; sorting nexin 22; sorting nexin-22
Gene Symbols SNX22
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX22. Recombinant protein control fragment (Product #RP-98113).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79856
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.