Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX22 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 130621AP150UL

Description
The protein encoded by this gene is a sorting nexin that is found in the cytoplasm, where it interacts with membrane-bound phosphatidylinositol 3-phosphate. The encoded protein may play a role in intracellular trafficking. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.Specifications
SNX22 | |
Polyclonal | |
Unconjugated | |
SNX22 | |
SNX22, sorting nexin 22 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
382083, 79856 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96L94 | |
Snx22 | |
SNX22 Fusion Protein Ag3751 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction