Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX25 Monoclonal Antibody (3A8)
GREENER_CHOICE

Catalog No. PIMA524494
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524494 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524494 Supplier Invitrogen™ Supplier No. MA524494
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MYMERMDKRA LISFWESVEH LKNANKNEIP QLVGEIYQNF FVESKEISVE KSLYKEIQQC LVGNKGIEVF YKIQEDVYET LKDRYYPSFI VSDLYEKLLI KEEEKHASQM ISNKDEMGPR DEAGEEAVDD GTNQINEQAS FAVNKLRELN EKLEYKRQAL NSIQNAPKPD KKIVSKLKDE IILIEKERTD LQLHMARTDW WCENLGMWKA SITSGEVTEE NGEQLPCYFV MVSLQEVGGV ETKNWTVPRR LSEFQNLHRK LSECVPSLKK VQLPSLSKLP FKSIDQKFME KSKNQLNKFL QEETEEDSDL SDYGDDVDGR KDALAEPCFM LIGEIFELRG MFKWVRRTLI ALVQVTFGRT INKQIRDTVS WIFSEQMLVY YINIFRDAFW PNGKLAPPTT IRSKEQSQET KQRAQQKLLE NIPDMLQSLV GQQNARHGII KIFNALQETR ANKHLLYALM ELLLIELCPE LRVHLDQLKA GQV.

SNX25 (sorting nexin 25), also known as SBBI31 or MSTP043, is an 840 amino acid protein suggested to function in several stages of intracellular trafficking. A member of the sorting nexin family, SNX25 contains one PX (phox homology) domain, an RGS domain and one PXA domain. May be involved in several stages of intracellular trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX25
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3A8
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SNX25
Gene Accession No. Q9H3E2
Gene Alias AI661919; Gm1699; Gm173; MSTP043; Sbbi31; SNX25; sorting nexin 25; sorting nexin-25
Gene Symbols SNX25
Host Species Mouse
Immunogen SNX25 (AAH29868, 1 a.a. ~ 483 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83891
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.