Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX25 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30487220UL
Description
SNX25 Polyclonal specifically detects SNX25 in Human samples. It is validated for Western Blot.Specifications
SNX25 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
sorting nexin 25 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 230-530 of human SNX25 (NP_114159.2). | |
20 μL | |
Signal Transduction | |
83891 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction