Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNX26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556350
Description
Immunogen sequence: PPYLPRQQSD GSLLRSQRPM GTSRRGLRGP AQVSAQLRAG GGGRDAPEAA AQSPCSVPSQ VPTPGFFSPA PRECLPPFLG VPKP Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 75%.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Alternative splice variants encoding different isoforms have been identified in this gene.
Specifications
| SNX26 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP33 | |
| ARHGAP33; AW324378; FLJ39019; neurite outgrowth multiadaptor RhoGAP protein; NOMA-GAP; Rho GTPase activating protein 33; rho GTPase-activating protein 33; rho GTPase-activating protein 33; LOW QUALITY PROTEIN: rho GTPase-activating protein 33; rho-type GTPase-activating protein 33; Snx26; sorting nexin 26; sorting nexin-26; Tc10/CDC42 GTPase-activating protein; TCGAP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115703 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14559 | |
| ARHGAP33 | |
| Recombinant protein corresponding to Human SNX26. Recombinant protein control fragment (Product #RP-95545). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction