Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556351
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556351 Supplier Invitrogen™ Supplier No. PA556351
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSECVELFTE RPGPGLKADA DGPPCGIPAP QGISSLTSAV PRPRGKLAGL LRTFMRSRPS RQRLRQRGIL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Alternative splice variants encoding different isoforms have been identified in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX26
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGAP33
Gene Accession No. O14559
Gene Alias ARHGAP33; AW324378; FLJ39019; neurite outgrowth multiadaptor RhoGAP protein; NOMA-GAP; Rho GTPase activating protein 33; rho GTPase-activating protein 33; rho GTPase-activating protein 33; LOW QUALITY PROTEIN: rho GTPase-activating protein 33; rho-type GTPase-activating protein 33; Snx26; sorting nexin 26; sorting nexin-26; Tc10/CDC42 GTPase-activating protein; TCGAP
Gene Symbols ARHGAP33
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX26. Recombinant protein control fragment (Product #RP-95544).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115703
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.