Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570968 Supplier Invitrogen™ Supplier No. PA570968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Alternative splice variants encoding different isoforms have been identified in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX26
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ARHGAP33
Gene Accession No. O14559
Gene Alias ARHGAP33; AW324378; FLJ39019; neurite outgrowth multiadaptor RhoGAP protein; NOMA-GAP; Rho GTPase activating protein 33; rho GTPase-activating protein 33; rho GTPase-activating protein 33; LOW QUALITY PROTEIN: rho GTPase-activating protein 33; rho-type GTPase-activating protein 33; Snx26; sorting nexin 26; sorting nexin-26; Tc10/CDC42 GTPase-activating protein; TCGAP
Gene Symbols ARHGAP33
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human SNX26.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115703
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.