Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144741 Supplier Invitrogen™ Supplier No. PA5144741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSVPPHEADN LDPSDDSLGQ SFYDYTEKQA VPISVPRYKH VEQNGEKFVV YNVYMAGRQL CSKRYREFAI LHQNLK Antigen sequence identity: Mouse 99%; Rat 99%.

SNX27 encodes a member of the sorting nexin family, a diverse group of cytoplasmic and membrane-associated proteins involved in endocytosis of plasma membrane receptors and protein trafficking through these compartments. All members of this protein family contain a phosphoinositide binding domain (PX domain). A highly similar protein in mouse is responsible for the specific recruitment of an isoform of serotonin 5-hydroxytryptamine 4 receptor into early endosomes, suggesting the analogous role for the human protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX27
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX27
Gene Accession No. Q96L92
Gene Alias 5730552M22Rik; ESTM45; ESTM47; KIAA0488; MAP-responsive gene protein; methamphetamine (MAP) responsive transcript 1; methamphetamine-responsive transcript 1; methamphetamine-responsive transcript 1 protein; Mrt1; MY014; PDZ protein Mrt1; PDZ-protein Mrt1; R75405; RP11-98D18.12-005; SNX27; sorting nexin family member 27; sorting nexin-27
Gene Symbols SNX27
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 133-208.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81609
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.