Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX27 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987256
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7256 Supplier Bioss Supplier No. BS6424R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNX27 encodes a member of the sorting nexin family, a diverse group of cytoplasmic and membrane-associated proteins involved in endocytosis of plasma membrane receptors and protein trafficking through these compartments. All members of this protein family contain a phosphoinositide binding domain (PX domain). A highly similar protein in mouse is responsible for the specific recruitment of an isoform of serotonin 5-hydroxytryptamine 4 receptor into early endosomes, suggesting the analogous role for the human protein.

Specifications

Antigen SNX27
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SNX27
Gene Accession No. Q96L92
Gene Alias 5730552M22Rik; ESTM45; ESTM47; KIAA0488; MAP-responsive gene protein; methamphetamine (MAP) responsive transcript 1; methamphetamine-responsive transcript 1; Methamphetamine-responsive transcript 1 protein; Mrt1; My014; PDZ protein Mrt1; PDZ-protein Mrt1; R75405; RP11-98D18.12-005; Snx27; sorting nexin family member 27; Sorting nexin-27
Gene Symbols SNX27
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SNX27.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81609
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.