Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX29 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | SNX29 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1792620
![]() |
Novus Biologicals
NBP17926120UL |
20 μL |
Each for $158.00
|
|
|||||
NBP179261
![]() |
Novus Biologicals
NBP179261 |
100 μL |
Each for $487.50
|
|
|||||
Description
SNX29 Polyclonal specifically detects SNX29 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SNX29 | |
Polyclonal | |
Purified | |
RUO | |
A-388D4.1, FLJ00143, FLJ12363, FLJ40609, RUN domain containing 2A, sorting nexin 29 | |
SNX29 | |
IgG | |
Protein A purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
NP_115543 | |
92017 | |
Synthetic peptide directed towards the N terminal of human RUNDC2AThe immunogen for this antibody is RUNDC2A. Peptide sequence MSGSQNNDKRQFLLERLLDAVKQCQIRFGGRKEIASDSDSRVTCLCAQFE. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title