Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX29 Monoclonal Antibody (6D6)
GREENER_CHOICE

Catalog No. PIMA522730
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522730 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522730 Supplier Invitrogen™ Supplier No. MA522730
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MDEERSSMLP TMAAGPNSIL FAINIDNKDL NGQSKFAPTV SDLLKESTQN VTLLKESTQG VSSVFREITA SSAISILIKP EQETDPLPVV SRNVSADAKC KKERKKKKQV TNIISFDDEE DEQNSGDMFK KTPGAGESSE DNSDHSSVNI MSAFESPFGP NSNGSQSSNS WKIDSLSLNR EFGYQKLDVK SIDDEDVDEN EDDVYGNSSG RKHRGHSESP EKNGAHSVTQ AGVQWHDLGS LQPLPPGFR.

SNX29 also known as RUNDC2A, is a 375 amino protein that contains one RUN domain. RUNDC2A is encoded by a gene that maps to human chromosome 16p13.13, which encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX29
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6D6
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SNX29
Gene Accession No. Q8TEQ0, Q9D3S3
Gene Alias 4933437K13Rik; A-388D4.1; ENSMUSG00000079738; Gm11170; Gm1737; Gm930; RUN domain containing 2A; RUN domain-containing protein 2A; RUNDC2A; SNX29; SODD; sorting nexin 29; sorting nexin-29
Gene Symbols SNX29
Host Species Mouse
Immunogen RUNDC2A (AAH41583, 1 a.a. ~ 249 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74478, 92017
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.