Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX3 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$216.50 - $330.00
Specifications
Antigen | SNX3 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SNX3 Polyclonal antibody specifically detects SNX3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SNX3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, pH7.0 | |
8724 | |
Primary | |
Store at 4&drg;C short term. Aliquot and store at -20&drg;C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Grd19, MCOPS8, MGC17570, Protein SDP3, SDP3, sorting nexin 3, sorting nexin 3A, sorting nexin-3 | |
Produced in rabbits immunized with E. coli-derived Human SNX3 fragment | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title