Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553974
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553974 Supplier Invitrogen™ Supplier No. PA553974
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGGDSTPSPD LLMARSFGDK DLILPNGGTP AGTSSPASSS SLLNRLQLDD DIDGETRDLF VIVDDPKKHV CTMETY Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 99%.

SNX9, SNX18 and SNX30 constitute a separate subfamily of PX-BAR-containing sorting nexin (SNX) proteins. May be involved in several stages of intracellular trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX30
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX30
Gene Accession No. Q5VWJ9
Gene Alias 4732481H14Rik; ATG24A; C030041J06Rik; hypothetical protein LOC550496; novel PX domain containing protein; RGD1311367; Snx30; sorting nexin family member 30; sorting nexin-30; zgc:112424
Gene Symbols SNX30
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX30. Recombinant protein control fragment (Product #RP-93003).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401548
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.