Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559721 Supplier Invitrogen™ Supplier No. PA559721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YHTRIRDACL RADRVMRAHK CLADDYIPIS AALSSLGTQE VNQLRTSFLK LAELFERLRK LEG Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 86%.

SNX32 (sorting nexin-32), also known as SNX6B (sorting nexin-6B), is a 403 amino acid protein that contains one PX (phox homology) domain and belongs to the sorting nexin family. Existing as two alternatively spliced isoforms, SNX32 may be involved in several stages of intracellular trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX32
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX32
Gene Accession No. Q86XE0
Gene Alias B930037P14Rik; RGD1560591; SNX32; Snx6b; sortin nexin-6B; sorting nexin 32; sorting nexin 6B; sorting nexin-32; Sorting nexin-6B
Gene Symbols SNX32
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX32. Recombinant protein control fragment (Product #RP-97475).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254122
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.