Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559378
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559378 Supplier Invitrogen™ Supplier No. PA559378
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRSGVEAFIL GDVPMMAKIA ETYSIEMGPR GPQWKANPHP FACSVEDPTK QTK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

SNX33 (sorting nexin-33), also known as SH3PX3, SH3PXD3C or SNX30, is a 574 amino acid protein that interacts with ADAM15 and FAS-L. Belonging to the sorting nexin family, SNX33 contains one BAR domain, one PX (phox homology) domain and one SH3 domain. The gene that encodes SNX33 consists of over 14,000 bases and maps to human chromosome 15q24.2. Housing approximately 106 million base pairs and encoding more than 700 genes, chromosome 15 makes up about 3% of the human genome. Angelman and Prader-Willi syndromes are associated with loss of function or deletion of genes in the 15q11-q13 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX33
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX33
Gene Accession No. Q8WV41
Gene Alias SH3 and PX domain containing 3; SH3 and PX domain-containing protein 3; SH3PX3; SH3PXD3C; SNX30; SNX33; sorting nexin 33; sorting nexin-33
Gene Symbols SNX33
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX33. Recombinant protein control fragment (Product #RP-98138).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257364
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.