Learn More
Invitrogen™ SNX4 Monoclonal Antibody (OTI1B1)

Description
SNX4 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | SNX4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1B1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SNX4 |
| Gene Accession No. | O95219, Q91YJ2 |
| Gene Alias | 1810036H14Rik; AI037066; ATG24B; Snx4; sorting nexin 4; sorting nexin-4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.