Learn More
Invitrogen™ SNX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552188
Description
Immunogen sequence: LRVRARVADR LYGVYKVHGN YGRVFSEWSA IEKEMGDGLQ SAGHHMDVYA SSIDDILEDE EHYADQLKEY LFYAEALRAV CRKHELMQYD LEMAAQDLAS KKQQCEELVT GTVRTFSLKG MTTKLFGQET PEQREARIKV LEEQINEGE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
SNX2 and SNX4 share a high degree of amino acid similarity with SNX1, as they all contain a characteristic phox homology (PX) domain. These proteins are all partially associated with cellular membranes, and they, likewise, associate with EGF, PDGF and insulin receptor tyrosine kinases. These nexins are widely expressed and yet have various tissue distribution patterns. Additionally, the sorting nexins can associate with each other and with a variety of other cellular proteins, suggesting that they exist as part of multisubunit complexes. The related protein, SNX3, comprises a distinct subgroup of nexins that share less sequence similarity outside of the PX domain and have dramatically different binding affinities for the tyrosine kinase receptors.
Specifications
SNX4 | |
Polyclonal | |
Unconjugated | |
SNX4 | |
1810036H14Rik; AI037066; ATG24B; Snx4; sorting nexin 4; sorting nexin-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
360725, 69150, 8723 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95219, Q91YJ2 | |
SNX4 | |
Recombinant protein corresponding to Human SNX4. Recombinant protein control fragment (Product #RP-102402). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.