Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589038 Supplier Invitrogen™ Supplier No. PA589038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFLQRLSSHP VLSKDRNFHV FLEYDQDLSV RRKNTKEMFG GFFKSVVKSA DEVLFTGVKE VDDFFEQEKN FLINYYNRIK DSCVKADKMT RSHKNVADDY IHTAACLHSL ALEEPTVIKK YLLKVAELFE KLRKVEGRVS SDEDLKLTEL LRYYMLNIEA AKDLLYRRTK ALIDYENSNK ALDKARLKSK DVKLAEAHQQ ECCQKFEQLS ESAKEELINF KRKRVAAFRK NLIEMSELEI KHARNNVSLL QSCIDLFKNN; Positive Samples: HepG2, U-87MG, Mouse thymus, Mouse brain, Mouse kidney, Rat thymus; Cellular Location: Cell membrane, Cell projection, Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle membrane, Early endosome, Early endosome membrane, Endosome, Peripheral membrane protein, phagocytic cup, ruffle.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein binds to fanconi anemia complementation group A protein, but its function is unknown. This gene results in two transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SNX5
Gene Accession No. B1H267, Q9D8U8, Q9Y5X3
Gene Alias 0910001N05Rik; 1810032P22Rik; AU019504; D2Ertd52e; FLJ10931; id:ibd5091; RP11-504H3.2; Snx5; snx5 protein; sorting nexin 5; sorting nexin-5; wu:fa66c10; wu:fi28h04; zgc:55769
Gene Symbols SNX5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 145-404 of human SNX5 (NP_0552411).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27131, 296199, 69178
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.