Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557945
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557945 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557945 Supplier Invitrogen™ Supplier No. PA557945
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IADHPTLTFN EDFKIFLTAQ AWELSSHKKQ GPGLLSRMGQ TVRAVASSMR GVKNRPEEFM EMNNFIELFS QKINLIDKIS QRIYKEE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region like some family members, and its function is unknown. This gene results in two transcript variants encoding two distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX7
Gene Accession No. Q9UNH6
Gene Alias 2510028H01Rik; SNX7; sorting nexin 7; sorting nexin-7
Gene Symbols SNX7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX7. Recombinant protein control fragment (Product #RP-94395).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51375
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.