Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110411
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110411 Supplier Invitrogen™ Supplier No. PA5110411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKFEDEPDLK DLFITVDEPE SHVTTIETFI TYRIITKTSR GEFDSSEFEV RRRYQDFLWL KGKLEEAHPT LIIPPLPEKF IVKGMVERFN DDFIETRRKA LHKFLNRIAD HPTLTFNEDF KIFLTAQAWE LSSHKKQGPG LLSRMGQTVR AVASSMRGVK NRPEEFMEMN NFIELFSQKI NLIDKISQRI YKEEREYFDE MKEYGPIHIL WSASEEDLVD TLKDVASCID RCCKATEKRM SGLSEA.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region like some family members, and its function is unknown. This gene results in two transcript variants encoding two distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SNX7
Gene Accession No. Q9CY18
Gene Alias 2510028H01Rik; SNX7; sorting nexin 7; sorting nexin-7
Gene Symbols SNX7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 85-330 of human SNX7 (NP_057060.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310815, 76561
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.